Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Covidien Monoject™ Angel Wing™ Blood Collection/Infusion Sets with Female Luer

Catalog No. 22042907
Click to view available options
Needle Gauge:
19 G
21 G
23 G
25 G
Color:
Blue
Cream
Green
Orange
Length (English) Tube:
3 in.
12 in.
This item is not returnable. View return policy
This item is not returnable. View return policy

Used for routine venipunctures and infusion applications

  • One-handed activation of safety mechanism prevents accidental needlesticks
  • Textured wings identify bevel and provide ease of control
  • Flexible tubing prevents coiling
  • Latex-free to minimize potential allergic reaction
  • 0.75 in. (1.9cm) needle length

Specifications

Product Type Winged Blood Collection Set
Color Blue
Needle Gauge 23 G
Length (English) Needle 0.75 in.
Length (English) Tube 12 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.