Learn More
Covidien Sterile Midstream Urine Collection Systems
Click to view available options
Includes:
Castille Soap Wipes
Protective Ring, Castille Wipes
Description
Includes
Castille soap towelette
- The moisture-resistant packaging has a fiber-free seal that guards against contamination before opening
- Clear packaging allows nursing to describe contents and the user to view instructions
- Sequentially packed contents guide user through the procedure
- Available with protective ring to guard against contamination of sample and for ease of handling
Specifications
Specifications
Product Type | Midstream Specimen Catch Kit |
Sterility | Sterile |
Includes | Castille Soap Wipes |
Volume (English) | 0.03 gal. |
Volume (Metric) | 118 mL |