Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ COX-1, Rabbit Polyclonal Antibody

Catalog No. RB10687R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RB10687R1 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RB10687R1 Supplier Epredia™ Supplier No. RB10687R1

Ensure accurate and reproducible results in immunohistochemistry experiments with Epredia™ COX-1, Rabbit Polyclonal Antibody.

Cyclooxygenases 1 and 2 (COX-1, COX-2) are enzymes involved in the conversion of arichidonate to prostaglandins. COX-1 and COX-2 are very similar in structure and function, though they vary in expression. COX-1 is normally expressed in most cell types, whereas COX-2 expression is at low levels unless induced by hormonal stimuli. In some breast cancer and bladder carcinoma studies, COX-1 was expressed in the stroma cells adjacent to the tumors but not in tumor cells.

Host Species: Rabbit

Species Reactivity: Human. Others not tested.

Epitope: C-terminal

Immunogen: Synthetic peptide corresponding to C-terminal of human COX-1

Molecular Weight: 72 kDa

Positive Control: Esophagus

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen COX-1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Immunogen Synthetic peptide corresponding to C-terminal of human COX-1
Purification Method Esophagus
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Prediluted
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.