Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COX-2 Antibody (4A9), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | COX-2 |
---|---|
Clone | 4A9 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
COX-2 Monoclonal antibody specifically detects COX-2 in Human samples. It is validated for ELISASpecifications
COX-2 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
COX-2, COX2cyclooxygenase 2b, cyclooxygenase-2, EC 1.14.99, EC 1.14.99.1, GRIPGHS, hCox-2, PGG/HS, PGH synthase 2, PGHS-2, PHS II, PHS-2, prostaglandin G/H synthase 2, prostaglandin G/H synthase and cyclooxygenase, Prostaglandin H2 synthase 2, Prostaglandin-endoperoxide synthase 2, prostaglandin-endoperoxide synthase 2 (prostaglandin G/H synthase andcyclooxygenase) | |
A synthesized peptide derived from Human COX-2 [UniProt P35354] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
4A9 | |
ELISA | |
Unconjugated | |
Rabbit | |
Cancer, Cell Biology, Cellular Markers, Cytokine Research, Hypoxia, Lipid and Metabolism, Phospho Specific, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
5743 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title