Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX-2 Mouse anti-Human, Clone: COX2/1941, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen COX-2
Clone COX2/1941
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, ELISA 2-4 ug/ml
Applications Western Blot, ELISA
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121976
SDP
View Documents
Novus Biologicals
NBP30725120UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

COX-2 Monoclonal specifically detects COX-2 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

COX-2
0.2 mg/ml
Western Blot, ELISA
Unconjugated
Mouse
Cancer, Cell Biology, Cellular Markers, Cytokine Research, Hypoxia, Lipid and Metabolism, Phospho Specific, Signal Transduction
10 mM PBS with 0.05% BSA
5743
IgG2b κ
Protein A or G purified
COX2/1941
Western Blot 1-2 ug/ml, ELISA 2-4 ug/ml
Monoclonal
Purified
RUO
Human
COX-2, COX2cyclooxygenase 2b, cyclooxygenase-2, EC 1.14.99, EC 1.14.99.1, GRIPGHS, hCox-2, PGG/HS, PGH synthase 2, PGHS-2, PHS II, PHS-2, prostaglandin G/H synthase 2, prostaglandin G/H synthase and cyclooxygenase, Prostaglandin H2 synthase 2, Prostaglandin-endoperoxide synthase 2, prostaglandin-endoperoxide synthase 2 (prostaglandin G/H synthase andcyclooxygenase)
Recombinant human COX-2 protein fragment (around aa 442-572) (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.