Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COX10 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297977AF594
Description
COX10 Polyclonal specifically detects COX10 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
COX10 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
COX10 (yeast) homolog, cytochrome c oxidase assembly protein (heme A:farnesyltransferase), COX10 homolog, cytochrome c oxidase assembly protein, heme A:farnesyltransferase (yeast), cytochrome c oxidase assembly protein, cytochrome c oxidase subunit X, EC 2.5.1.-, heme A: farnesyltransferase, Heme O synthase, protoheme IX farnesyltransferase, mitochondrial | |
Produced in rabbits immunized with E. coli-derived Human COX10 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
1352 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction