Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560591
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560591 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560591 Supplier Invitrogen™ Supplier No. PA560591
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DKPVIGISTY NIVPFEAGQY FNKIQCFCFE EQRLNPQEEV DMPVFFYIDP EFAEDPRMIK VDLITLSYTF FEAKEGHKLP VPG Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.

Cytochrome c oxidase (COX) is the terminal enzyme in the electron transfer chain, functioning as a transmembrane proton pump that builds an electrochemical gradient with chemical energy from the reduction of O2. Cytochrome c oxidase assembly protein COX11 is an intracellular mitochondrial membrane protein necessary for the construction of an active COX complex. COX11 contains a single transmembrane helix downstream of the N-terminal, mitochondrial targeting sequence and a C-terminal Cu(I)-binding domain. The assembly of COX requires the delivery of metal cofactors. Along with COX12 and SCO1/2, COX11 acts as a metal ion chaperone necessary for copper insertion into CuA and CuB redox-active copper centers of COX in eukaryotes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX11
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COX11
Gene Accession No. Q9Y6N1
Gene Alias 2010004I09Rik; Cox11; COX11 cytochrome c oxidase copper chaperone; COX11 homolog, cytochrome c oxidase assembly protein; COX11, cytochrome c oxidase copper chaperone; COX11P; cytochrome c oxidase assembly homolog 11; cytochrome c oxidase assembly protein 11; cytochrome c oxidase assembly protein COX11, mitochondrial; cytochrome c oxidase subunit 11
Gene Symbols COX11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COX11. Recombinant protein control fragment (Product #RP-100211).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1353
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.