Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COX11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104519
Description
Antibody detects endogenous levels of total COX11.
Cytochrome c oxidase (COX) is the terminal enzyme in the electron transfer chain, functioning as a transmembrane proton pump that builds an electrochemical gradient with chemical energy from the reduction of O2. Cytochrome c oxidase assembly protein COX11 is an intracellular mitochondrial membrane protein necessary for the construction of an active COX complex. COX11 contains a single transmembrane helix downstream of the N-terminal, mitochondrial targeting sequence and a C-terminal Cu(I)-binding domain. The assembly of COX requires the delivery of metal cofactors. Along with COX12 and SCO1/2, COX11 acts as a metal ion chaperone necessary for copper insertion into CuA and CuB redox-active copper centers of COX in eukaryotes.
Specifications
COX11 | |
Polyclonal | |
Unconjugated | |
COX11 | |
2010004I09Rik; Cox11; COX11 cytochrome c oxidase copper chaperone; COX11 homolog, cytochrome c oxidase assembly protein; COX11, cytochrome c oxidase copper chaperone; COX11P; cytochrome c oxidase assembly homolog 11; cytochrome c oxidase assembly protein 11; cytochrome c oxidase assembly protein COX11, mitochondrial; cytochrome c oxidase subunit 11 | |
Rabbit | |
Affinity chromatography | |
RUO | |
1353, 690300, 69802 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q6P8I6, Q9Y6N1 | |
COX11 | |
A synthesized peptide derived from human COX11(Accession Q9Y6N1), corresponding to amino acid residues S60-S110. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction