Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560781
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560781 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560781 Supplier Invitrogen™ Supplier No. PA560781
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VRVYHYFQWR RAQRQAAEEQ KTSGIM Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 73%.

Core component of the MITRAC (mitochondrial translation regulation assembly intermediate of cytochrome c oxidase complex) complex, that regulates cytochrome c oxidase assembly. Requires for coordination of the early steps of cytochrome c oxidase assembly with the synthesis of MT-CO1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX14
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COX14
Gene Accession No. Q96I36
Gene Alias C12orf62; COX14; COX14 cytochrome c oxidase assembly factor; COX14 cytochrome c oxidase assembly homolog; COX14, cytochrome c oxidase assembly factor; cytochrome c oxidase assembly homolog 14; cytochrome c oxidase assembly protein COX14
Gene Symbols COX14
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COX14. Recombinant protein control fragment (Product #RP-97854).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84987
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.