Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX15 Monoclonal Antibody (2D2)
GREENER_CHOICE

Catalog No. PIMA518633
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518633 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518633 Supplier Invitrogen™ Supplier No. MA518633
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: LTESGLSMVD WHLIKEMKPP TSQEEWEAEF QRYQQFPEFK ILNHDMTLTE FKFIWYMEYS H.

The cytochrome c oxidase (COX) family of proteins function as the final electron donor in the respiratory chain to drive a proton gradient across the inner mitochondrial membrane, ultimately resulting in the production of water. The mammalian COX apoenzyme is a dimer, with each monomer consisting of 13 subunits, some of which are mitochondrial and some of which are nuclear. COX15 (cytochrome c oxidase assembly protein COX15 homolog) is a 410 amino acid multi-pass mitochondrial membrane protein belonging to the COX15/ctaA family. Existing as two alternatively spliced isoforms, COX15 is predominantly expressed in tissues containing high rates of oxidative phosphorylation including brain, heart and muscle. COX15 defects are the cause of a neurodegenerative disorder known as Leigh syndrome and cytochrome c oxidase deficiency (COX deficiency).
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX15
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2D2
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene COX15
Gene Accession No. Q7KZN9
Gene Alias 2900026G05Rik; CEMCOX2; COX15; COX15 cytochrome c oxidase assembly homolog; COX15 homolog, cytochrome c oxidase assembly protein; COX15, cytochrome c oxidase assembly homolog; cytochrome c oxidase assembly homolog 15; cytochrome c oxidase assembly homolog 15 (yeast); cytochrome c oxidase assembly protein 15; cytochrome c oxidase assembly protein COX15 homolog; cytochrome c oxidase subunit 15
Gene Symbols COX15
Host Species Mouse
Immunogen COX15 (NP_510870, 92 a.a. ~ 152 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1355
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.