Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589973
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589973 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589973 Supplier Invitrogen™ Supplier No. PA589973
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YSHRMWGRLV GLVYILPAAY FWRKGWLSRG MKGRVLALCG LVCFQGLLGW YMVKSGLEEK SDSHDIPRVS QYRLAAHLGS ALVLYCASLW TSLSLLLPPH K; Positive Samples: U-251MG, A-431, Jurkat, Mouse heart, Mouse kidney, Mouse lung, Rat testis; Cellular Location: Mitochondrion membrane, Multi-pass membrane protein.

The cytochrome c oxidase (COX) family of proteins function as the final electron donor in the respiratory chain to drive a proton gradient across the inner mitochondrial membrane, ultimately resulting in the production of water. The mammalian COX apoenzyme is a dimer, with each monomer consisting of 13 subunits, some of which are mitochondrial and some of which are nuclear. COX15 (cytochrome c oxidase assembly protein COX15 homolog) is a 410 amino acid multi-pass mitochondrial membrane protein belonging to the COX15/ctaA family. Existing as two alternatively spliced isoforms, COX15 is predominantly expressed in tissues containing high rates of oxidative phosphorylation including brain, heart and muscle. COX15 defects are the cause of a neurodegenerative disorder known as Leigh syndrome and cytochrome c oxidase deficiency (COX deficiency).
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX15
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene COX15
Gene Accession No. Q7KZN9, Q8BJ03
Gene Alias 2900026G05Rik; CEMCOX2; COX15; COX15 cytochrome c oxidase assembly homolog; COX15 homolog, cytochrome c oxidase assembly protein; COX15, cytochrome c oxidase assembly homolog; cytochrome c oxidase assembly homolog 15; cytochrome c oxidase assembly homolog 15 (yeast); cytochrome c oxidase assembly protein 15; cytochrome c oxidase assembly protein COX15 homolog; cytochrome c oxidase subunit 15
Gene Symbols COX15
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 150-250 of human COX15 (NP_5108701).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1355, 226139, 309391
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.