Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564046
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564046 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564046 Supplier Invitrogen™ Supplier No. PA564046
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IKDSKFDDWK NIRGPRPWED PDLLQGRNPE SLKTKT Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.

This protein is encoded by COX16, corresponding to the previously uncharacterized open reading frame YJL003w of the yeast genome. COX16 was identified in studies of COX-deficient mutants previously assigned to complementation group G22 of a collection of yeast pet mutants. To determine its location, Cox16p was tagged with a Myc epitope at the C terminus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX16
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cox16
Gene Accession No. Q9P0S2
Gene Alias 1810020G14Rik; 1810055I05Rik; BB388670; C14orf112; COX16; COX16 cytochrome c oxidase assembly homolog; COX16, cytochrome c oxidase assembly homolog; cytochrome c oxidase assembly factor; cytochrome c oxidase assembly protein 16; cytochrome c oxidase assembly protein COX16 homolog, mitochondrial; hCOX16; HSPC203; PTD019
Gene Symbols Cox16
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COX16. Recombinant protein control fragment (Product #RP-102011).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51241
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.