Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX17 Monoclonal Antibody (OTI2D7), TrueMAB™, OriGene
SDP

Catalog No. 502543805
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3805 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3805 Supplier OriGene Supplier No. CF812118
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

COX17 Monoclonal antibody specifically detects COX17 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen COX17
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2D7
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene Hspa1l
Gene Accession No. P16627, P34931, P55063
Gene Alias AI037035; Cox17; COX17 cytochrome c oxidase assembly homolog; COX17 cytochrome c oxidase copper chaperone; COX17 homolog, cytochrome c oxidase assembly protein; COX17, cytochrome c oxidase copper chaperone; Cox17a; cytochrome c oxidase 17 copper chaperone; cytochrome c oxidase assembly homolog 17; cytochrome c oxidase assembly protein 17; cytochrome c oxidase copper chaperone; cytochrome c oxidase subunit XVII assembly protein homolog; cytochrome c oxidase, subunit XVII assembly protein homolog; human homolog of yeast mitochondrial copper recruitment
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 400-641 of human HSPA1L (NP_005518) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15482, 24963, 3305
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.