Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561317
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561317 Supplier Invitrogen™ Supplier No. PA561317
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PTLPVWAVAP VSAVHANGWY EALAASSPVR VAEEVLLGVH Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 68%.

COX18 encodes a cytochrome c oxidase (COX)-assembly protein. The S. cerevisiae Cox18 protein catalyzes the insertion of the Cox2 (MTCO2; MIM 516040) C-terminal tail into the mitochondrial inner membrane, an intermediate step in the assembly of complex IV of the mitochondrial respiratory chain. Required for the insertion of integral membrane proteins into the mitochondrial inner membrane. Essential for the activity and assembly of cytochrome c oxidase. Plays a central role in the translocation and export of the C-terminal part of the COX2 protein into the mitochondrial intermembrane space.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX18
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cox18
Gene Accession No. Q8N8Q8
Gene Alias BC038311; COX18; COX18 cytochrome c oxidase assembly factor; COX18 cytochrome c oxidase assembly homolog; COX18, cytochrome c oxidase assembly factor; COX18Hs; cytochrome c oxidase assembly factor COX18; cytochrome c oxidase assembly homolog 18; cytochrome c oxidase assembly protein 18; Cytochrome c oxidase assembly protein COX18, mitochondrial; mitochondrial inner membrane protein COX18; Oxa1l2; RGD1559547
Gene Symbols Cox18
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COX18. Recombinant protein control fragment (Product #RP-100829).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285521
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.