Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554355
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554355 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554355 Supplier Invitrogen™ Supplier No. PA554355
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NFGTKSFQPR PPDKGSFPLD HLGECKSFKE KFMKCLHNNN FENALCRKES KEYLECRMER KLMLQEPLEK LGFGD Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 84%.

COX19 encodes a cytochrome c oxidase (COX)-assembly protein. The S. cerevisiae Cox19 protein may play a role in metal transport to the mitochondrial intermembrane space and assembly of complex IV of the mitochondrial respiratory chain (Sacconi et al., 2005 [PubMed 16212937]).[supplied by OMIM, Mar 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX19
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COX19
Gene Accession No. Q49B96
Gene Alias 1810074D02Rik; 2810437L13Rik; COX19; COX19 cytochrome c oxidase assembly factor; COX19 cytochrome c oxidase assembly homolog; COX19, cytochrome c oxidase assembly factor; cytochrome c oxidase assembly homolog 19; cytochrome c oxidase assembly protein 19; cytochrome c oxidase assembly protein COX19; hCOX19
Gene Symbols COX19
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COX19. Recombinant protein control fragment (Product #RP-92838).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90639
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.