Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX2 Recombinant Rabbit Monoclonal Antibody (10B2)
GREENER_CHOICE

Catalog No. PIMA538394
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538394 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538394 Supplier Invitrogen™ Supplier No. MA538394
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody has been tested in direct-ELISA.

COX2 converts arachidonate to prostaglandin H2 (PGH2), a committed step in prostanoid synthesis, including production of inflammatory prostaglandins. The conversion of arachidonate to prostaglandin H2 is a 2 step reaction: a cyclooxygenase (COX) reaction which converts arachidonate to prostaglandin G2 (PGG2) and a peroxidase reaction in which PGG2 is reduced to prostaglandin H2 (PGH2). It is constitutively expressed in some tissues in physiological conditions, such as the endothelium, kidney and brain, and is up-regulated under pathological conditions, such as in cancer and inflammation (in contrast to the iso-enzyme PTGS1, which is expressed ubiquitously). Up-regulation of COX2 is also associated with increased cell adhesion, phenotypic changes, resistance to apoptosis and tumor angiogenesis. In cancer cells, COX2 is a key step in the production of prostaglandin E2 (PGE2), which plays important roles in modulating motility, proliferation and resistance to apoptosis. COX2 is naturally inhibited by calcitriol (the active form of Vitamin D). Glucocorticoids chronically trans-repress PTGS2 gene activity in vivo in part by interfering with transcription initiation and elongation. COX2 is a target of NSAID such as aspirin, which can reduce pain and swelling from inflammation driven by COX2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX2
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 10B2
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 150mM NaCl and 0.02% sodium azide; pH 7.4
Gene PTGS2
Gene Accession No. P35354
Gene Alias COII; COX2; Cox-2; COXII; cyclooxygenase; cyclooxygenase 2; cyclooxygenase 2b; cyclooxygenase-2; Cytochrome c oxidase polypeptide II; Cytochrome c oxidase subunit 2; cytochrome c oxidase subunit II; glucocorticoid-regulated inflammatory cyclooxygenase; Gripghs; hCox-2; Macrophage activation-associated marker protein P71/73; mitochondrially encoded cytochrome c oxidase II; MTCO2; MT-CO2; PES-2; PGG/HS; PGH synthase 2; Pghs2; PGHS-2; Pghs-b; PHS II; PHS-2; prostaglandin G/H synthase 2; Prostaglandin H2 synthase 2; prostaglandin-endoperoxide synthase 2; prostaglandin-endoperoxide synthase 2 (prostaglandin G/H synthase and cyclooxygenase); Ptgs2; Tis10; TIS10 protein
Gene Symbols PTGS2
Host Species Rabbit
Immunogen A synthesized peptide derived from human COX2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5743
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.