Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX2 Mouse anti-Human, Clone: 12C4F12, Abnova™

Catalog No. 89035421 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-421 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-421 Supplier Abnova Corporation Supplier No. MAB14016

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against native human COX2.

  • High specificity
  • High reactivity

Specifications

Antigen COX2
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 12C4F12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native human COX2.
Dilution Immunocytochemistry (5-10 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In HEPES (0.02% sodium azide).
Gene Accession No. P00403
Gene Alias MTCO2
Gene Symbols COX2
Host Species Mouse
Immunogen Native purified human COX2.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4513
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.