Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561018
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561018 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561018 Supplier Invitrogen™ Supplier No. PA561018
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YNYAKQRIQE RIAREEIKKK ILYEGTHLDP ERKHNGSSSN Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 78%.

COX20, also name as FAM36A, belongs to the COX20 family. Mutations in COX20 are a novel cause of recessively inherited.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX20
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COX20
Gene Accession No. Q5RI15
Gene Alias COX20; COX20 Cox2 chaperone homolog; COX20 cytochrome c oxidase assembly factor; COX20, cytochrome c oxidase assembly factor; Cytochrome c oxidase assembly protein COX20, mitochondrial; cytochrome c oxidase protein 20 homolog; FAM36A; family with sequence similarity 36, member A
Gene Symbols COX20
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COX20. Recombinant protein control fragment (Product #RP-99826).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116228
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.