Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109988
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109988 Supplier Invitrogen™ Supplier No. PA5109988
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTHQSHAYHM VKPSPWPLTG ALSALLMTSG LAMWFHFHSM TLLMLGLLTN TLTMYQWWRD VTRESTYQGH HTPPVQKGLR YGMILFITSE VFFFAGFFWA.

COX3 is a multi-pass membrane protein. It belongs to the cytochrome c oxidase subunit 3 family. Defects in COX3 are a cause of Leber hereditary optic neuropathy (LHON) and cytochrome c oxidase deficiency (COX deficiency). Defects in MT-CO3 are also found in mitochondrial encephalomyopathy with lactic acidosis and stroke-like episodes (MELAS) syndrome and recurrent myoglobinuria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.06 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene COX3
Gene Accession No. P00414, P00416, P05505
Gene Alias ACI60_gp01; Co3; Coiii; COX3; COXIII; cytchrome c oxidase sububnit 3; Cytochrome c oxidase polypeptide III; Cytochrome c oxidase subunit 3; cytochrome c oxidase subunit III; mitochondrially encoded cytochrome c oxidase III; Mtco3; mt-Co3; OXI2; Q0275
Gene Symbols COX3, MT-CO3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MT-CO3 (YP_0030240321).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139542, 17710, 26204, 4514
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.