Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COX4 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11039115R
Description
COX4 Polyclonal specifically detects COX4 in Human, Mouse, Rat, Porcine, Bovine, Drosophila, Insect, Opossum, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
COX4 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, KnockDown Validated | |
P13073 | |
COX4I1 | |
A synthetic peptide made to an internal region of human COX IV isoform 1 (within residues 1-100). [Swiss-Prot# P13073] | |
0.1 mL | |
Cancer, Cellular Markers, Hypoxia, Lipid and Metabolism | |
1327 | |
Human, Mouse, Rat, Pig, Bovine, Drosophila, Insect, Opossum, Primate | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
COX IV-1, COX4-1, COX4COXIV, Cytochrome c oxidase polypeptide IV, cytochrome c oxidase subunit 4 isoform 1, mitochondrial, cytochrome c oxidase subunit IV isoform 1cytochrome c oxidase subunit IV, MGC72016 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, mouse, bovine, primate and rat. Immunogen sequence has 76% homology to Zebrafish. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction