Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse COX4 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems MAB6980SP
This item is not returnable.
View return policy
Description
COX4 Monoclonal specifically detects COX4 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| COX4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q96KJ9 | |
| COX4I2 | |
| E.coli-derived recombinant human COX4-I2 Asn39-Lys171 Accession # Q96KJ9 | |
| 25 μg | |
| Primary | |
| Detects human COX4-I2 in direct ELISAs. In Western blots, 100% cross-reactivity with recombinant human (rh) COX4-I1 and no cross-reactivity with rhCOX-1 or rhCOX-2 is observed. | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 673803 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| COX IV-1, COX4-1, COX4-2, COX4B, COX4COXIV, COX4I1, COX4I2, COX4L2, COXIV-2, Cytochrome c oxidase polypeptide IV, cytochrome c oxidase subunit 4 isoform 1, mitochondrial, cytochrome c oxidase subunit IV isoform 1cytochrome c oxidase subunit IV, MGC72016 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1327 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction