Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COX4, p Ser58 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP229532
Description
COX4 Polyclonal specifically detects COX4 in Mouse, Rat samples. It is validated for Western Blot.Specifications
COX4 (p Ser58) | |
Polyclonal | |
Western Blot 1:1000 | |
P19783 | |
COX4I1 | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser58 conjugated to KLH. Accession # P19783 | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Specific for the ∼17k COXIV-1 protein phosphorylated at Ser58. Immunolabeling is blocked by the phospho-peptide used as antigen but not by the corresponding dephospho-peptide. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
COX IV-1, COX4-1, COX4COXIV, Cytochrome c oxidase polypeptide IV, cytochrome c oxidase subunit 4 isoform 1, mitochondrial, cytochrome c oxidase subunit IV isoform 1cytochrome c oxidase subunit IV, MGC72016 | |
Rabbit | |
17 kDa | |
0.1 mL | |
Cancer, Cellular Markers, Hypoxia, Lipid and Metabolism | |
1327 | |
Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction