Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX4I2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101306
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101306 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101306 Supplier Invitrogen™ Supplier No. PA5101306
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total COX42.

Component of the cytochrome c oxidase, the last enzyme in the mitochondrial electron transport chain which drives oxidative phosphorylation. The respiratory chain contains 3 multisubunit complexes succinate dehydrogenase (complex II, CII), ubiquinol-cytochrome c oxidoreductase (cytochrome b-c1 complex, complex III, CIII) and cytochrome c oxidase (complex IV, CIV), that cooperate to transfer electrons derived from NADH and succinate to molecular oxygen, creating an electrochemical gradient over the inner membrane that drives transmembrane transport and the ATP synthase. Cytochrome c oxidase is the component of the respiratory chain that catalyzes the reduction of oxygen to water. Electrons originating from reduced cytochrome c in the intermembrane space (IMS) are transferred via the dinuclear copper A center (CU(A)) of subunit 2 and heme A of subunbit 1 to the active site in subunit 1, a binuclear center (BNC) formed by heme A3 and copper B (CU(B)). The BNC reduces molecular oxygen to 2 water molecules using 4 electrons from cytochrome c in the IMS and 4 protons from the mitochondrial matrix. [Uniprot].
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX4I2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene COX4I2
Gene Accession No. Q91W29, Q91Y94, Q96KJ9
Gene Alias AL024441; COX; cox iv; COX IV-1; Cox IV-2; cox4; COX4-1; Cox4-2; Cox4a; Cox4b; cox4i1; cox4i1.S; Cox4i2; COX4L2; COXIV; COXIV-1; CoxIV-2; Cytochrome c oxidase polypeptide IV; cytochrome c oxidase subunit 4 isoform 1, mitochondrial; Cytochrome c oxidase subunit 4 isoform 2, mitochondrial; cytochrome c oxidase subunit 4I1; cytochrome c oxidase subunit 4I2; cytochrome c oxidase subunit IV; cytochrome c oxidase subunit IV isoform 1; cytochrome c oxidase subunit IV isoform 1 precursor; cytochrome c oxidase subunit IV isoform 1 S homeolog; cytochrome c oxidase subunit IV isoform 2; cytochrome c oxidase subunit IV isoform 2 (lung); cytochrome c oxidase subunit IV precursor EC 1.9.3.1; cytochrome c oxidase subunit IV-like 2; cytochrome c oxidase, subunit 4a; cytochrome c oxidase, subunit 4b; cytochrome c oxidase, subunit IV; cytochrome c oxidase, subunit IVa; cytochrome c oxidase, subunit IVb; cytochrome c oxydase subunit 4; dJ857M17.2; FLJ23483; hypothetical protein MGC73355; IV-1; mg:bb02d03; MGC72016; XELAEV_18024950mg; zgc:110058; zgc:73355
Gene Symbols COX4I2
Host Species Rabbit
Immunogen A synthesized peptide derived from human COX4I2(Accession Q96KJ9), corresponding to amino acid residues Y41-T91.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84682, 84683, 84701
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.