Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COX4 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11039115AF532
Description
COX4 Polyclonal specifically detects COX4 in Human, Mouse, Rat, Porcine, Bovine, Drosophila, Insect, Opossum, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| COX4 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, KnockDown Validated | |
| COX IV-1, COX4-1, COX4COXIV, Cytochrome c oxidase polypeptide IV, cytochrome c oxidase subunit 4 isoform 1, mitochondrial, cytochrome c oxidase subunit IV isoform 1cytochrome c oxidase subunit IV, MGC72016 | |
| Rabbit | |
| 0.1 mL | |
| 1327 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| COX4I1 | |
| A synthetic peptide made to an internal region of human COX IV isoform 1 (within residues 1-100). [Swiss-ProtNo. P13073] | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine, Drosophila, Insect, Opossum, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction