Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COX4I1 Rabbit anti-Human, Unconjugated, Clone: AIC-3, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB19914
Description
COX4I1 Monoclonal antibody specifically detects COX4I1 in Human samples. It is validated for IP, ICC, WB-Ce, IHC-P, IF, Flow Cyt.Specifications
COX4I1 | |
Monoclonal | |
Unconjugated | |
COX4I1 | |
Rabbit | |
100 μg | |
1327 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
AIC-3 | |
cytochrome c oxidase subunit IV isoform 1 | |
COX4, COXIV, MGC72016 | |
A synthetic peptide corresponding to human COX4I1 | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction