Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COX4NB Antibody (OTI1D6), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271595IR
Description
COX4NB Monoclonal specifically detects COX4NB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
COX4NB | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EMC8 | |
Full length recombinant protein of human COX4NB (NP_006058) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1D6 | |
Western Blot, Immunohistochemistry | |
C16orf2, C16orf4, chromosome 16 open reading frame 2, COX4 neighbor, COX4AL, FAM158Bchromosome 16 open reading frame 4, neighbor of COX4, NOC4family with sequence similarity 158, member B, Protein FAM158B | |
Mouse | |
Affinity Purified | |
Core ESC Like Genes, Stem Cell Markers | |
10328.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction