Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX4NB Monoclonal Antibody (OTI5G1), TrueMAB™, OriGene
SDP

Catalog No. 501676939
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-6939 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-6939 Supplier OriGene Supplier No. TA507066
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Expressed in liver, pancreas, heart, lung, kidney, brain, skeletal muscle, and placenta, Expression levels are highest in pancreas and moderate in heart, skeletal muscle, and placenta.

Specifications

Antigen COX4NB
Applications Western Blot
Classification Monoclonal
Clone OTI5G1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene EMC8
Gene Accession No. O43402
Gene Alias C16orf2, C16orf4, COX4NB, FAM158B, NOC4
Gene Symbols EMC8
Host Species Mouse
Immunogen Full length human recombit protein of human COX4NB produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10328
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.