Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX4NB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561984
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561984 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561984 Supplier Invitrogen™ Supplier No. PA561984
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VKLTTQAYCK MVLHGAKYPH CAVNGLLVAE KQKPRKEHLP LGGPGAHHTL FVD Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.

EMC8 (ER Membrane Protein Complex Subunit 8) is a Protein Coding gene. An important paralog of this gene is EMC9.[GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX4NB
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Emc8
Gene Accession No. O43402
Gene Alias C16orf2; C16orf4; COX4 neighbor; COX4AL; COX4NB; Emc8; ER membrane protein complex subunit 8; FAM158B; family with sequence similarity 158, member B; neighbor of COX4; NOC4; Protein FAM158B
Gene Symbols Emc8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COX4NB. Recombinant protein control fragment (Product #RP-98556).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10328
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.