Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COX5A Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306072R
Description
COX5A Polyclonal specifically detects COX5A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
COX5A | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
COX, COX-VA, Cytochrome c oxidase polypeptide Va, cytochrome c oxidase polypeptide, mitochondrial, cytochrome c oxidase subunit 5A, mitochondrial, cytochrome c oxidase subunit Va, mitochondrial cytochrome c oxidase subunit Va, VA | |
Produced in rabbits immunized with E. coli-derived Human COX5A fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
9377 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction