Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX5A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104367
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104367 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104367 Supplier Invitrogen™ Supplier No. PA5104367
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total COX5A.

Cytochrome c oxidase (COX) is the terminal enzyme of the mitochondrial respiratory chain. It is a multi-subunit enzyme complex that couples the transfer of electrons from cytochrome c to molecular oxygen and contributes to a proton electrochemical gradient across the inner mitochondrial membrane. The complex consists of 13 mitochondrial- and nuclear-encoded subunits. The mitochondrially-encoded subunits perform the electron transfer of proton pumping activities. The functions of the nuclear-encoded subunits are unknown but they may play a role in the regulation and assembly of the complex. This gene encodes the nuclear-encoded subunit Va of the human mitochondrial respiratory chain enzyme. A pseudogene COX5AP1 has been found in chromosome 14q22.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX5A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene COX5A
Gene Accession No. P11240, P12787, P20674
Gene Alias AA959768; CcOX; COX; Cox5a; COX5A protein; COX-VA; cytochrome c oxidase polypeptide Va; cytochrome c oxidase polypeptide, mitochondrial; cytochrome c oxidase subunit 5A; cytochrome c oxidase subunit 5A, mitochondrial; cytochrome c oxidase subunit Va; cytochrome c oxidase, subunit Va; mitochondrial cytochrome c oxidase subunit 5a; mitochondrial cytochrome c oxidase subunit Va; VA
Gene Symbols COX5A
Host Species Rabbit
Immunogen A synthesized peptide derived from human COX5A(Accession P20674), corresponding to amino acid residues L23-G73.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12858, 252934, 9377
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.