Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COX5b Antibody (021), Alexa Fluor™ 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290312AF488
Description
COX5b Monoclonal specifically detects COX5b in Human samples. It is validated for Western Blot, ELISA.Specifications
COX5b | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
21 | |
Western Blot, ELISA | |
COXVB, Cytochrome c oxidase polypeptide Vb, cytochrome c oxidase polypeptide VB, mitochondrial, cytochrome c oxidase subunit 5B, mitochondrial, cytochrome c oxidase subunit Vb | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human COX5b (P10606; Ala32-His129). | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
1329 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction