Learn More
COX5B Monoclonal Antibody (16H12H9)
COX5B Monoclonal Antibody (16H12H9)
Supplier: LIFE TECHNOLOGIES 459110

Description
COX5B Monoclonal Antibody for Western Blot, Flow

Specifications
COX5B | |
Monoclonal | |
1 mg/mL | |
HEPES buffered saline with 0.02% sodium azide | |
P00428, P10606, P12075, P19536 | |
COX5B | |
Bovine Complex IV (native holoenzyme protein, purified from heart). | |
100 μg | |
Primary | |
Human, Mouse, Rat, Bovine | |
Antibody | |
IgG2b κ |
Flow Cytometry, Western Blot | |
16H12H9 | |
Unconjugated | |
COX5B | |
COX5B; COXVB; cytochrome c oxidase polypeptide Vb; cytochrome c oxidase polypeptide VB, mitochondrial; cytochrome c oxidase polypeptide VIa; cytochrome c oxidase precursor (EC 1.9.3.1); cytochrome c oxidase subunit 5B; cytochrome c oxidase subunit 5B, mitochondrial; cytochrome c oxidase subunit Vb; cytochrome c oxidase subunit VIA* | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
12859, 1329, 287012, 94194 | |
4°C, do not freeze | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.