Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COX6A1 Monoclonal Antibody (OTI3F1), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA501397S


Description
COX6A1 Monoclonal antibody specifically detects COX6A1 in Human samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
COX6A1 | |
Monoclonal | |
0.52 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P12074 | |
Mouse | |
Affinity chromatography | |
RUO | |
1337 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunocytochemistry | |
OTI3F1 | |
Unconjugated | |
Cox6a1 | |
CMTRID; COX VIa-L; COX6A; Cox6a1; Cox6al; COXVIA2; Cytochrome c oxidase polypeptide VIa-liver; cytochrome c oxidase subunit 6A1; cytochrome c oxidase subunit 6A1, mitochondrial; Cytochrome c oxidase subunit SSG; cytochrome c oxidase subunit VIa; Cytochrome c oxidase subunit VIa (liver); cytochrome C oxidase subunit VIa homolog; cytochrome c oxidase subunit VIa polypeptide 1; cytochrome c oxidase subunit VIA-liver; cytochrome c oxidase, subunit VIa, polypeptide 1; cytochrome oxidase subunit VIa; cytochrome-c oxidase; ns:zf-e250; subunit VIaL (liver-type); Unknown (protein for MGC:140155); VIA L; VIaL; zgc:103782 | |
Full length human recombinant protein of human COX2H6 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction