Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX6A1 Monoclonal Antibody (OTI6H8), TrueMAB™, OriGene
SDP

Catalog No. 502542317
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2317 30 μL
50-167-2541 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2317 Supplier OriGene Supplier No. TA501398S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

COX6A1 Monoclonal antibody specifically detects COX6A1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry

Specifications

Antigen COX6A1
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone OTI6H8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Cox6a1
Gene Accession No. P12074
Gene Alias CMTRID; COX VIa-L; COX6A; Cox6a1; Cox6al; COXVIA2; Cytochrome c oxidase polypeptide VIa-liver; cytochrome c oxidase subunit 6A1; cytochrome c oxidase subunit 6A1, mitochondrial; Cytochrome c oxidase subunit SSG; cytochrome c oxidase subunit VIa; Cytochrome c oxidase subunit VIa (liver); cytochrome C oxidase subunit VIa homolog; cytochrome c oxidase subunit VIa polypeptide 1; cytochrome c oxidase subunit VIA-liver; cytochrome c oxidase, subunit VIa, polypeptide 1; cytochrome oxidase subunit VIa; cytochrome-c oxidase; ns:zf-e250; subunit VIaL (liver-type); Unknown (protein for MGC:140155); VIA L; VIaL; zgc:103782
Host Species Mouse
Immunogen Full length human recombinant protein of human COX6A1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1337
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.