Learn More
COX6A1 Mouse anti-Human, Clone: OTI2H6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501396

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This protein is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.Specifications
COX6A1 | |
Monoclonal | |
Unconjugated | |
COX6A1 | |
CMTRID, COX6A, COX6AL | |
Mouse | |
Affinity Chromatography | |
RUO | |
1337 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI2H6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P12074 | |
COX6A1 | |
Full length human recombit protein of human COX2H6 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.