Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COX6A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596256
Description
Immunogen sequence: SSGAHGEEGS ARMWKTLTFF VALPGVAVSM LNVYLKSHHG EHERPEFIAY PHLRIRTKPF PWGDGNHTLF HNPHVNPLPT GYEDE; Positive Samples: A-549, MCF7, Mouse kidney, Mouse heart, Mouse liver, Rat liver, Rat brain; Cellular Location: Mitochondrion inner membrane.
This protein is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.
Specifications
COX6A1 | |
Polyclonal | |
Unconjugated | |
Cox6a1 | |
CMTRID; COX VIa-L; COX6A; Cox6a1; Cox6al; COXVIA2; Cytochrome c oxidase polypeptide VIa-liver; cytochrome c oxidase subunit 6A1; cytochrome c oxidase subunit 6A1, mitochondrial; Cytochrome c oxidase subunit SSG; cytochrome c oxidase subunit VIa; Cytochrome c oxidase subunit VIa (liver); cytochrome C oxidase subunit VIa homolog; cytochrome c oxidase subunit VIa polypeptide 1; cytochrome c oxidase subunit VIA-liver; cytochrome c oxidase, subunit VIa, polypeptide 1; cytochrome oxidase subunit VIa; cytochrome-c oxidase; ns:zf-e250; subunit VIaL (liver-type); Unknown (protein for MGC:140155); VIA L; VIaL; zgc:103782 | |
Rabbit | |
Affinity chromatography | |
RUO | |
12861, 1337, 25282 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.79 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P10818, P12074, P43024 | |
Cox6a1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 25-109 of human COX6A1 (NP_0043642). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction