Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX6B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109577
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109577 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109577 Supplier Invitrogen™ Supplier No. PA5109577
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAEDMETKIK NYKTAPFDSR FPNQNQTRNC WQNYLDFHRC QKAMTAKGGD ISVCEWYQRV YQSLCPTSWV TDWDEQRAEG TFPGKI.

Connects the two COX monomers into the physiological dimeric form.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX6B1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene COX6B1
Gene Accession No. P14854, P56391
Gene Alias 2010000G05Rik; COX VIb-1; Cox6b; COX6B1; COXG; COXVIb1; cytochrome c oxidase subunit 6B1; cytochrome c oxidase subunit VIb; Cytochrome c oxidase subunit VIb isoform 1; cytochrome c oxidase subunit VIb polypeptide 1 (ubiquitous); cytochrome c oxidase, subunit VIb polypeptide 1; LOC688869; similar to cytochrome c oxidase, subunit VIb polypeptide 1
Gene Symbols COX6B1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-86 of human COX6B1 (NP_001854.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110323, 1340
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.