Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX6B1 Recombinant Rabbit Monoclonal Antibody (7T0J5)
GREENER_CHOICE

Catalog No. PIMA542770
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542770 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542770 Supplier Invitrogen™ Supplier No. MA542770
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: 293T, SH-SY5Y. The target is usually found in the following locations: Mitochondrion intermembrane space. Immunogen sequence: MAEDMETKIK NYKTAPFDSR FPNQNQTRNC WQNYLDFHRC QKAMTAKGGD ISVCEWYQRV YQSLCPTSWV TDWDEQRAEG TFPGKI.

Connects the two COX monomers into the physiological dimeric form.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX6B1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 7T0J5
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene COX6B1
Gene Accession No. P14854
Gene Alias 2010000G05Rik; COX VIb-1; Cox6b; COX6B1; COXG; COXVIb1; cytochrome c oxidase subunit 6B1; cytochrome c oxidase subunit VIb; Cytochrome c oxidase subunit VIb isoform 1; cytochrome c oxidase subunit VIb polypeptide 1 (ubiquitous); cytochrome c oxidase, subunit VIb polypeptide 1; LOC688869; similar to cytochrome c oxidase, subunit VIb polypeptide 1
Gene Symbols COX6B1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-86 of human COX6B1 (P14854).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1340
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.