Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COX6B2 Antibody (OTI1F5), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272379MFV450
Description
COX6B2 Monoclonal antibody specifically detects COX6B2 in Human samples. It is validated for Flow CytometrySpecifications
COX6B2 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
125965 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
OTI1F5 | |
Flow Cytometry | |
Cancer/testis antigen 59, COX VIb-2, COXVIB2, CT59MGC119094, cytochrome c oxidase subunit 6B2, Cytochrome c oxidase subunit VIb isoform 2, cytochrome c oxidase subunit VIb polypeptide 2 (testis), cytochrome c oxidase subunit VIb, testes specific, cytochrome c oxidase subunit VIb, testes-specific, Cytochrome c oxidase subunit VIb, testis-specific isoform, FLJ32865, FLJ46422 | |
Full length human recombinant protein of human COX6B2 (NP_653214) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction