Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX6B2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550213
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550213 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550213 Supplier Invitrogen™ Supplier No. PA550213
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total COX6B2 protein.

The cytochrome c oxidase (COX) family of proteins function as the final electron donor in the respiratory chain to drive a proton gradient across the inner mitochondrial membrane, ultimately resulting in the production of water. The mammalian COX apoenzyme is a dimer, with each monomer consisting of 13 subunits, some of which are mitochondrial and some of which are nuclear. Localized to the intermembrane space, COX6b2 (Cytochrome c oxidase subunit 6B2), also known as Cytochrome c oxidase subunit VIb isoform 2 and Cancer/testis antigen 59, is a 88 amino acid mitochondrial protein that is responsible for joining the two COX monomers to form the COX dimer. COX6b2 is specifically expressed in testis and is found to be upregulated in certain cancer cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX6B2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene COX6B2
Gene Accession No. Q6YFQ2
Gene Alias 1700067P11Rik; BC048670; Cancer/testis antigen 59; COX VIb-2; Cox6b2; COXVIB2; CT59; Cytochrome c oxidase subunit 6B2; Cytochrome c oxidase subunit VIb isoform 2; cytochrome c oxidase subunit VIb polypeptide 2; cytochrome c oxidase subunit VIb polypeptide 2 (testis); cytochrome c oxidase subunit VIb testes-specific isoform; cytochrome c oxidase subunit VIb, testes-specific; cytochrome c oxidase subunit VIb, testis-specific isoform
Gene Symbols COX6B2
Host Species Rabbit
Immunogen Fusion protein corresponding to residues near the N terminal of human cytochrome c oxidase subunit VIb polypeptide 2 (testis).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125965
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.