Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX6C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549921
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549921 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549921 Supplier Invitrogen™ Supplier No. PA549921
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total COX6C protein.

Cytochrome c oxidase, the terminal enzyme of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. It is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may be involved in the regulation and assembly of the complex. This nuclear gene encodes subunit VIc, which has 77% amino acid sequence identity with mouse subunit VIc. This gene is up-regulated in prostate cancer cells. A pseudogene has been found on chromosomes 16p12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX6C
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene COX6C
Gene Accession No. P09669, P11951, Q9CPQ1
Gene Alias COVIc; Cox6c; Cox6c2; COX-VIc; cytochrome c oxidase polypeptide VIc; Cytochrome c oxidase polypeptide VIc-2; cytochrome c oxidase subunit 6C; Cytochrome c oxidase subunit 6C-2; cytochrome c oxidase subunit VIc; cytochrome c oxidase subunit VIc preprotein; cytochrome c oxidase, subunit VIc; cytochrome oxidase subunit VIc; EC 1.9.3.1
Gene Symbols COX6C
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human COX6C.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12864, 1345, 54322
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.