Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COX6C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106557
Description
Antibody detects endogenous levels of total COX6C.
Cytochrome c oxidase, the terminal enzyme of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. It is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may be involved in the regulation and assembly of the complex. This nuclear gene encodes subunit VIc, which has 77% amino acid sequence identity with mouse subunit VIc. This gene is up-regulated in prostate cancer cells. A pseudogene has been found on chromosomes 16p12.
Specifications
COX6C | |
Polyclonal | |
Unconjugated | |
COX6C | |
COVIc; Cox6c; Cox6c2; COX-VIc; cytochrome c oxidase polypeptide VIc; Cytochrome c oxidase polypeptide VIc-2; cytochrome c oxidase subunit 6C; Cytochrome c oxidase subunit 6C-2; cytochrome c oxidase subunit VIc; cytochrome c oxidase subunit VIc preprotein; cytochrome c oxidase, subunit VIc; cytochrome oxidase subunit VIc; EC 1.9.3.1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
12864, 1345, 54322 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P09669, P11951, Q9CPQ1 | |
COX6C | |
A synthesized peptide derived from human COX6C(Accession P09669), corresponding to amino acid residues R20-I70. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction