Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COX7A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570677
Description
This target displays homology in the following species: Cow: 79%; Dog: 79%; Horse: 79%; Human: 100%; Mouse: 75%; Pig: 79%; Rabbit: 77%; Sheep: 79%; Zebrafish: 79%.
This protein is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.
Specifications
| COX7A1 | |
| Polyclonal | |
| Unconjugated | |
| COX7A1 | |
| COX; COX7A; Cox7a1; Cox7ah; COX7AM; cytochrome c oxidase polypeptide VIIa-heart; cytochrome c oxidase polypeptide VIIa-muscle/heart; cytochrome c oxidase subunit 7A1; Cytochrome c oxidase subunit 7A1, mitochondrial; cytochrome c oxidase subunit VIIa 1; cytochrome c oxidase subunit VIIa heart/muscle isoform; cytochrome c oxidase subunit VIIa polypeptide 1 (muscle); Cytochrome c oxidase subunit VIIa-H; Cytochrome c oxidase subunit VIIa-heart; cytochrome c oxidase subunit VIIa-M; Cytochrome c oxidase subunit VIIa-muscle; cytochrome c oxidase subunit VIIIc; cytochrome c oxidase, subunit VIIa 1; cytochrome-c oxidase subunit VIIa; VIIa-M; VIIIC; zgc:153177 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1346 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P24310 | |
| COX7A1 | |
| synthetic peptide directed towards the middle region of human COX7A1 (aa 7-56). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction