Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX7A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599811
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599811 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599811 Supplier Invitrogen™ Supplier No. PA599811
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total COX7A1.

This protein is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX7A1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene COX7A1
Gene Accession No. P24310, P56392
Gene Alias COX; COX7A; Cox7a1; Cox7ah; COX7AM; cytochrome c oxidase polypeptide VIIa-heart; cytochrome c oxidase polypeptide VIIa-muscle/heart; cytochrome c oxidase subunit 7A1; Cytochrome c oxidase subunit 7A1, mitochondrial; cytochrome c oxidase subunit VIIa 1; cytochrome c oxidase subunit VIIa heart/muscle isoform; cytochrome c oxidase subunit VIIa polypeptide 1 (muscle); Cytochrome c oxidase subunit VIIa-H; Cytochrome c oxidase subunit VIIa-heart; cytochrome c oxidase subunit VIIa-M; Cytochrome c oxidase subunit VIIa-muscle; cytochrome c oxidase subunit VIIIc; cytochrome c oxidase, subunit VIIa 1; cytochrome-c oxidase subunit VIIa; VIIa-M; VIIIC; zgc:153177
Gene Symbols COX7A1
Host Species Rabbit
Immunogen A synthesized peptide derived from human COX7A1(Accession P24310), corresponding to amino acid residues Q2-L52.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100910772, 12865, 1346
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.