Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX7A2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566237
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566237 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566237 Supplier Invitrogen™ Supplier No. PA566237
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YKFSGFTQKLA GAWASEAYSP QGLKPVVSTE APPIIFATPT KLTSDSTVYD YAGKNKVPEL QKFFQKADGV PVY Highest antigen sequence identity to the following orthologs - mouse 85%, rat 58%.

The cytochrome c oxidase (COX) family of proteins function as the final electron donor in the respiratory chain to drive a proton gradient across the inner mitochondrial membrane, ultimately resulting in the production of water. The mammalian COX apoenzyme is a dimer, with each monomer consisting of 13 subunits, some of which are mitochondrial and some of which are nuclear. COX7a2 (cytochrome c oxidase subunit VIIa polypeptide 2), also known as COX7AL or COX7AL1, is an 83 amino acid protein that localizes to the inner mitochondrial membrane and exists as a component of the COX complex, playing an important role in electron transport. COX7a2L (cytochrome c oxidase subunit 7A-related protein), also known as COX7AR or COX7RP, is an inner mitochondrial membrane protein that consists of 114 amino acids and is induced by estrogen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX7A2L
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cox7a2l
Gene Accession No. O14548
Gene Alias COX7A2L; COX7AR; COX7a-related protein; COX7RP; cytochrome c oxidase subunit 7A2 like; cytochrome c oxidase subunit 7A-related protein, mitochondrial; cytochrome c oxidase subunit VIIa polypeptide 2 like; cytochrome c oxidase subunit VIIa polypeptide 2-like; cytochrome c oxidase subunit VIIa-related protein; cytochrome c oxidase subunit VII-related protein; EB1; estrogen receptor binding CpG island; Scaf1; SIG81; SIG-81; Silg81; Silica-induced gene 81 protein; supercomplex assembly factor I
Gene Symbols Cox7a2l
Host Species Rabbit
Immunogen Recombinant Human COX7A2L. Recombinant protein control fragment (Product #RP-106919).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9167
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.