Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX7A2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599808
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599808 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599808 Supplier Thermo Scientific Supplier No. PA599808
Add to Cart
Add to Cart

COX7A2L Polyclonal Antibody

COX7A2L Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen COX7A2L
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Cox7a2l
Gene Accession No. O14548, Q61387
Gene Alias COX7A2L; COX7AR; COX7a-related protein; COX7RP; cytochrome c oxidase subunit 7A2 like; cytochrome c oxidase subunit 7A-related protein, mitochondrial; cytochrome c oxidase subunit VIIa polypeptide 2 like; cytochrome c oxidase subunit VIIa polypeptide 2-like; cytochrome c oxidase subunit VIIa-related protein; cytochrome c oxidase subunit VII-related protein; EB1; estrogen receptor binding CpG island; Scaf1; SIG81; SIG-81; Silg81; Silica-induced gene 81 protein; supercomplex assembly factor I
Gene Symbols Cox7a2l
Host Species Rabbit
Immunogen A synthesized peptide derived from human COX7A2L(Accession O14548), corresponding to amino acid residues P35-M85.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20463, 298762, 9167
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.