Learn More
Invitrogen™ COX7A2 Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 85%; Dog: 85%; Guinea Pig: 85%; Horse: 85%; Human: 100%; Mouse: 77%; Rabbit: 85%; Rat: 77%; Sheep: 85%; Zebrafish: 75%.
Specifications
Specifications
| Antigen | COX7A2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | COX7A2P2 |
| Gene Accession No. | O60397, P14406 |
| Gene Alias | Cox7a2; Cox7a3; COX7AL; COX7AL1; COX7AL2; COXVIIAL; COXVIIa-L; cytochrome c oxidase polypeptide 7A2, mitochondrial; cytochrome c oxidase polypeptide VIIa-liver/heart; cytochrome c oxidase subunit 7A2; cytochrome c oxidase subunit 7A2, mitochondrial; cytochrome c oxidase subunit VIIa 2; cytochrome c oxidase subunit VIIa 3; cytochrome c oxidase subunit VIIa polypeptide 2; cytochrome c oxidase subunit VIIa polypeptide 2 (liver); cytochrome c oxidase subunit VIIaL; cytochrome c oxidase subunit VIIa-L; Cytochrome c oxidase subunit VIIa-liver/heart; cytochrome c oxidase, subunit 7a 3; cytochrome c oxidase, subunit VIIa 2; hepatic cytochrome-c oxidase chain VIIa; VIIAL |
| Gene Symbols | COX7A2, COX7A2P2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.