Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX7A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570743
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570743 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570743 Supplier Invitrogen™ Supplier No. PA570743
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 92%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 77%; Rabbit: 79%; Rat: 83%.

This protein is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX7A2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene COX7A2P2
Gene Accession No. O60397, P14406
Gene Alias Cox7a2; Cox7a3; COX7AL; COX7AL1; COX7AL2; COXVIIAL; COXVIIa-L; cytochrome c oxidase polypeptide 7A2, mitochondrial; cytochrome c oxidase polypeptide VIIa-liver/heart; cytochrome c oxidase subunit 7A2; cytochrome c oxidase subunit 7A2, mitochondrial; cytochrome c oxidase subunit VIIa 2; cytochrome c oxidase subunit VIIa 3; cytochrome c oxidase subunit VIIa polypeptide 2; cytochrome c oxidase subunit VIIa polypeptide 2 (liver); cytochrome c oxidase subunit VIIaL; cytochrome c oxidase subunit VIIa-L; Cytochrome c oxidase subunit VIIa-liver/heart; cytochrome c oxidase, subunit 7a 3; cytochrome c oxidase, subunit VIIa 2; hepatic cytochrome-c oxidase chain VIIa; VIIAL
Gene Symbols COX7A2, COX7A2P2
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human COX7A3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1347, 1348
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.