Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX7B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543756
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543756 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543756 Supplier Invitrogen™ Supplier No. PA543756
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MFPLVKSALN RLQVRSIQQT MARQSHQKRT PDFHDKYGNA VLASGATFCI Sequence homology: Human: 100%; Mouse: 92%; Pig: 77%; Rabbit: 92%; Rat: 92%.

Cytochrome c oxidase (COX), the terminal component of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. This component is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may function in the regulation and assembly of the complex. This nuclear gene encodes subunit VIIb, which is highly similar to bovine COX VIIb protein and is found in all tissues. This gene may have several pseudogenes on chromosomes 1, 2, 20 and 22, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX7B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene COX7B
Gene Accession No. P24311
Gene Alias 1100001F07Rik; 1110004F07Rik; APLCC; C80563; COX7B; Cytochrome c oxidase polypeptide VIIb; cytochrome c oxidase subunit 7B; cytochrome c oxidase subunit 7B, mitochondrial; cytochrome c oxidase subunit VIIb; cytochrome-c oxidase chain VIIb; LSDMCA2
Gene Symbols COX7B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human COX7B (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1349
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.