Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX7B2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103404
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103404 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103404 Supplier Invitrogen™ Supplier No. PA5103404
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Cytochrome c Oxidase 7B2.

The cytochrome c oxidase (COX) family of proteins function as the final electron donor in the respiratory chain to drive a proton gradient across the inner mitochondrial membrane, ultimately resulting in the production of water and ATP. The mammalian COX apoenzyme is a dimer, with each monomer consisting of 13 subunits, some of which are mitochondrial and some of which are nuclear. COX7b (cytochrome c oxidase subunit VIIb polypeptide) and COX7b2 (cytochrome c oxidase subunit VIIb polypeptide 2) are 80 and 81 amino acid proteins, respectively, which exist as components of the COX complex, therefore playing an important role in electron transport. A rare polymorphism in the COX7b2 gene at codon 26 may be linked to nasopharyngeal carcinoma (NPC), the most common head and neck cancer in southern China.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX7B2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene COX7B2
Gene Accession No. Q8TF08
Gene Alias 4930503B16Rik; COX7B2; Cytochrome c oxidase polypeptide VIIb2; cytochrome c oxidase subunit 7B2; cytochrome c oxidase subunit 7B2, mitochondrial; cytochrome c oxidase subunit VIIb2
Gene Symbols COX7B2
Host Species Rabbit
Immunogen A synthesized peptide derived from human COX7B2(Accession Q8TF08), corresponding to amino acid residues P4-W54.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170712, 78174
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.